Learn More
SI Analytics™ 10 mL Exchangeable Unit
Product Code. 13485898 Shop All SI Analytics Products
Change view
Click to view available options
Volume (Metric):
10 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
13485898 10 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13485898 Supplier SI Analytics™ Supplier No. 285220310
SI Analytics™ 10 mL Exchangeable Unit is an accessory for TitroLine™ and Titronic™ models.
  • With integrated chip for reagent data and brown glass bottle for titrant
  • Includes GL 45 and S 40-bottle adapter
TRUSTED_SUSTAINABILITY
Specifications
Type WA 10
Includes Integrated chip, glass bottle, bottle adapter, drip tubes, and titration itp
Volume (Metric) 10 mL