missing translation for 'onlineSavingsMsg'
Learn More

10-Undecenoic acid, 99%

Product Code. p-7024450
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1000g
250g
This item is not returnable. View return policy

Product Code. 11489416

Brand: Thermo Scientific Alfa Aesar A19122.30

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 112-38-9 | C11H20O2 | 184.279 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
10-Undecenoic acid is used for the preparation of active pharmaceutical ingredients, cosmetics, perfumes, antidandruff shampoos and antimicrobial powders. It is used for the treatment of skin problems and antifungal agent. It is involved in the production of nylon-11, methyl undecylenate, undecylenic aldehyde and undecylenic alcohol. Further, it is used as musk in perfumes and aromas.

Solubility
Soluble in alcohol, chloroform, diethyl ether, benzene and fixed or volatile oils. Insoluble in hot water and cold water.

Notes
Incompatible with bases, oxidizing agents and reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-38-9
Molecular Formula C11H20O2
Molecular Weight (g/mol) 184.279
MDL Number MFCD00004442
InChI Key FRPZMMHWLSIFAZ-UHFFFAOYSA-N
Synonym 10-undecenoic acid, undecylenic acid, desenex, 10-hendecenoic acid, renselin, declid, sevinon, 10-undecylenic acid, 9-undecylenic acid, undecyl-10-enic acid
PubChem CID 5634
ChEBI CHEBI:35045
IUPAC Name undec-10-enoic acid
SMILES C=CCCCCCCCCC(=O)O

Specifications

Melting Point 22°C to 25°C
Density 0.91
Boiling Point 126°C to 128°C (1 mmHg)
Flash Point 160°C (320°F)
Refractive Index 1.4493
Quantity 250 g
Beilstein 1762631
Merck Index 14,9848
Solubility Information Soluble in alcohol,chloroform,diethyl ether,benzene and fixed or volatile oils. Insoluble in hot water and cold water.
Formula Weight 184.28
Percent Purity 99%
Chemical Name or Material 10-Undecenoic acid
Show More Show Less
compliance-icons
Product Identifier
  • 10-Undecenoic acid
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : 10-Undecenoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.