missing translation for 'onlineSavingsMsg'
Learn More

100% High Cetane Diesel Fuel, VHG™

Product Code. 17960806
Change view
Click to view available options
Quantity:
20 mL
100 mL
500 mL
Unit Size:
100mL
20mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17960806 500 mL 500mL
17911316 100 mL 100mL
17913936 20 mL 20mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17960806 Supplier VHG Labs Supplier No. VHGBDBLENDBLK500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

100% High Cetane Diesel Fuel
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material B100 Biodiesel
Analyte or Component Names B100 Biodiesel
Shipping Condition Room Temperature
Solvent or Matrix High Cetane Diesel Fuel
Quantity 500 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.