missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TdB Labs AB 70 kDa Lysine-dextran, Positively Charged Dextran for Biomedical Applications
Click to view available options
Quantity:
10 mg
100 mg
1 g
Unit Size:
100mg
10mg
1g
Description
- Product Overview: Lysine-dextran is a positively charged dextran modified with lysine groups, designed for biomedical applications including cellular uptake studies, surface conjugation, and enhanced fixation during imaging. Its positive charge supports interaction with negatively charged biomolecules and surfaces.
- Features:
- Positively Charged: Introduced lysine groups provide available amine functionalities and enhance binding.
- Water-Soluble and Biocompatible: Ready for biological use in aqueous environments.
- Applications:
- Biomedical delivery system modeling.
- Surface conjugation and crosslinking studies.
- Enhanced fixation for live and fixed-cell imaging workflows.
- Studies involving charged macromolecule behavior.
Specifications
Specifications
| Format | Vial |
| Product Type | Reagent |
| Quantity | 100 mg |
| Form | Crystalline Powder |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction