Learn More
Gonotec™ 100 mOsmol/kg Calibration Standard
Description
- Calibration standard 100 mOsmol/kg
- Can be used as an alternative to the calibration standards with 300 mOsmol/kg or 850 mOsmol/kg
- Can be used as a further calibration point for the Osmomat 030 with the option 3P
Specifications
Specifications
| For Use With (Equipment) | Osmomat 030 Freezing Point Osmometer |