missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Integrin alpha V beta 3 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems MAB12192-100
This item is not returnable.
View return policy
Description
Integrin alpha V beta 3 Monoclonal specifically detects Integrin alpha V beta 3 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.
Specifications
| Integrin alpha V beta 3 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| antigen identified by monoclonal L230, CD51, CD51 antigen, integrin alpha-V, integrin, alpha V (vitronectin receptor, alpha polypeptide, antigen CD51), MSK8, Vitronectin receptor subunit alpha, VNRADKFZp686A08142 | |
| Chinese Hamster Ovary cell line CHO-derived human Integrin alpha V beta 3, Human Integrin alpha V (Phe31-Val992) and Human Integrin beta 3 (Gly27-Asp718), Accession # NP_002201 and AAA52589 | |
| 100 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG |
| Flow Cytometry | |
| 2549B | |
| Flow Cytometry 0.25 ug/10^6 cells | |
| NP_002201 and AAA52589 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 3685 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction