Learn More
Thermo Scientific™ 1000mL BIOS Swinging Bucket Rotors
Product Code. p-6582261 Shop All Thermo Scientific Products
Click to view available options
Capacity:
1000 mL
6 x 1000 mL
8 x 1000 mL
Type:
Bucket
Cover
Swinging Bucket Rotor
Includes:
Cover for Round Bucket (set of 2)
Rotor, Round Buckets, 1000 mL Bottles
Round Bucket (Set of2)
Unit Size:
Each
Pack of 2
Product Code. 15540194
Brand: Thermo Scientific™ 75003961
Efficiently and reliably perform your bioprocessing methods with the Thermo Scientific™ 6 x 1000mL or 8 x 1000mL swinging bucket rotors.
The Thermo Scientific™ 1000mL BIOS swinging bucket rotors are designed for use with the Thermo Scientific™ Sorvall™ BIOS 16 bioprocessing centrifuge. These rotors, which are provided complete with buckets and bottles, have been engineered to simplify operation of the centrifuge while maximizing productivity.
Achieve efficient and reliable sample processing with the following features:
  • 8 x 1000mL (8L) capacity at 5,400 max rpm (8,500 x g max RCF) operation;
  • 6 x 1000mL (6L) capacity at 4,600 max rpm (7,117 x g max RCF) operation
  • Thermo Scientific™ Eco-Spin™ windshield provides energy savings up to 64% when compared with non-windshielded rotors
  • Compatible with Thermo Scientific™ Auto-Lid™ and Auto-ID™ enabling instant lid opening, storage and rotor identification that saves time and protects the integrity of your samples
  • 2-year warranty
Upgrade to Thermo Scientific™ Dura-Coat™ buckets with nickel-coating to improve protection against corrosion due to moisture, chemicals or alkaline solutions.
TRUSTED_SUSTAINABILITY
Specifications
Type Swinging Bucket Rotor
Capacity 6 x 1000 mL
Max. RCF 8,500 x g
Max. Speed 5,400 rpm
For Use With (Equipment) Thermo Scientific Sorvall BIOS 16 Centrifuge
Includes Rotor, Round Buckets, 1000 mL Bottles