Learn More
Vanillin Propylene Glycol Acetal, TRC
Click to view available options
Quantity:
100 g
Unit Size:
100g
Specifications
Specifications
| Chemical Name or Material | Vanillin Propylene Glycol Acetal |
| CAS | 68527-74-2 |
| Molecular Formula | C11H14O4 |
| Quantity | 100 g |
| Synonym | Vanillin 1,2-propylene glycol acetal, 2-Methoxy-4-(4-methyl-1,3-dioxolan-2-yl)phenol |
| InChI Formula | InChI=1S/C11H14O4/c1-7-6-14-11(15-7)8-3-4-9(12)10(5-8)13-2/h3-5,7,11-12H,6H2,1-2H3 |
| SMILES | COc1cc(ccc1O)C2OCC(C)O2 |
| IUPAC Name | 2-methoxy-4-(4-methyl-1,3-dioxolan-2-yl)phenol |
| Molecular Weight (g/mol) | 210.23 |
| Formula Weight | 210.0892 |