missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Deoxy-2,2-difluoro-beta-D-erythro-pentofuranosyl)pyrimidin-2,4(1H,3H)-dione (2'-Deoxy-2',2'-difluorouridine), Mikromol™

Product Code. 30051037
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
This item is not returnable. View return policy

Product Code. 30051037

Brand: Mikromol MM1257.04

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

1-(2-Deoxy-2,2-difluoro-beta-D-erythro-pentofuranosyl)pyrimidin-2,4(1H,3H)-dione (2'-Deoxy-2',2'-difluorouridine)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2',2'-difluoro-2'-deoxyuridine
CAS 114248-23-6
Analyte or Component Names 2',2'-difluoro-2'-deoxyuridine
Shipping Condition Room Temperature
Molecular Formula C9 H10 F2 N2 O5
Certifications/Compliance ISO 9001
Synonym 2'-Deoxy-2',2'-difluorouridine, 1-(2-Deoxy-2,2-difluoro-beta-D-erythro-pentofuranosyl)pyrimidin-2,4(1H,3H)-dione, Gemcitabine Hydrochloride Impurity C, Gemcitabine Impurity C, Gemcitabine Impurity C
InChI Formula InChI=1S/C9H10F2N2O5/c10-9(11)6(16)4(3-14)18-7(9)13-2-1-5(15)12-8(13)17/h1-2,4,6-7,14,16H,3H2,(H,12,15,17)/t4-,6-,7-/m1/s1
SMILES OC[C@H]1O[C@@H](N2C=CC(=O)NC2=O)C(F)(F)[C@@H]1O
IUPAC Name 1-[(2R,4R,5R)-3,3-difluoro-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione
Molecular Weight (g/mol) 264.18
Quantity 100 mg
Formula Weight 264.0558 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.