missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(2-Hydroxyphenyl)-3-phenylpentan-1-one, Mikromol™
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
1-(2-Hydroxyphenyl)-3-phenylpentan-1-one
Specifications
Specifications
| Chemical Name or Material | 1-(2-Hydroxyphenyl)-3-phenylpentan-1-one |
| CAS | 93651-85-5 |
| Analyte or Component Names | 1-(2-Hydroxyphenyl)-3-phenylpentan-1-one |
| Shipping Condition | Room Temperature |
| Molecular Formula | C17 H18 O2 |
| Certifications/Compliance | ISO 17025 |
| Synonym | 2'-Hydroxy-3-phenylvalerophenone (7CI), 1-(2-hydroxyphenyl)-3-phenylpentan-1-one, 2-Hydroxy-3-phenylvalerophenon, 2-Hydroxy-beta-phenylvalerophenone |
| InChI Formula | InChI=1S/C17H18O2/c1-2-13(14-8-4-3-5-9-14)12-17(19)15-10-6-7-11-16(15)18/h3-11,13,18H,2,12H2,1H3 |
| SMILES | CCC(CC(=O)c1ccccc1O)c2ccccc2 |
| IUPAC Name | 1-(2-hydroxyphenyl)-3-phenylpentan-1-one |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction