missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(3,4-Dimethoxyphenyl)-2-methylpropan-1-one, Mikromol™
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
1-(3,4-Dimethoxyphenyl)-2-methylpropan-1-one
Specifications
Specifications
| Chemical Name or Material | 1-(3,4-Dimethoxyphenyl)-2-methylpropan-1-one |
| CAS | 14046-55-0 |
| Analyte or Component Names | 1-(3,4-Dimethoxyphenyl)-2-methylpropan-1-one |
| Shipping Condition | Room Temperature |
| Molecular Formula | C12 H16 O3 |
| Certifications/Compliance | ISO 17025 |
| Synonym | 1-(3,4-Dimethoxyphenyl)-2-methylpropan-1-one, Verapamil Hydrochloride Imp. L (EP), Verapamil Imp. L (EP), Verapamil Hydrochloride Impurity L, Verapamil Impurity L |
| InChI Formula | InChI=1S/C12H16O3/c1-8(2)12(13)9-5-6-10(14-3)11(7-9)15-4/h5-8H,1-4H3 |
| SMILES | COc1ccc(cc1OC)C(=O)C(C)C |
| IUPAC Name | 1-(3,4-dimethoxyphenyl)-2-methylpropan-1-one |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction