missing translation for 'onlineSavingsMsg'
Learn More
Learn More
10-[2-[(2RS)-1-Methylpiperidin-2-yl]ethyl]-2-(methylsulfanyl)-10H-phenothiazine 5-Oxide (Thioridazine 5-Sulfoxide), Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
Description
10-[2-[(2RS)-1-Methylpiperidin-2-yl]ethyl]-2-(methylsulfanyl)-10H-phenothiazine 5-Oxide (Thioridazine 5-Sulfoxide)
Specifications
Specifications
| Chemical Name or Material | Thioridazine 5-Sulfoxide |
| CAS | 5-8-7776 |
| Analyte or Component Names | Thioridazine 5-Sulfoxide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C21 H26 N2 O S2 |
| Certifications/Compliance | ISO 9001 |
| Synonym | 10-[2-[(2RS)-1-Methylpiperidin-2-yl]ethyl]-2-(methylsulfanyl)-10H-phenothiazine 5-oxide, 10-[2-(1-Methyl-2-piperidinyl)ethyl]-2-(methylthio)-10H-phenothiazine 5-oxide, Phenothiazine, 10-[2-(1-methyl-2-piperidyl)ethyl]-2-(methylthio)-, 5-oxide (7CI,8CI), Thioridazine 5-oxide, Thioridazine 5-sulfoxide, Thioridazine sulfoxide, Thioridazine Hydrochloride Impurity C, Thioridazine Impurity C |
| InChI Formula | InChI=1S/C21H26N2OS2/c1-22-13-6-5-7-16(22)12-14-23-18-8-3-4-9-20(18)26(24)21-11-10-17(25-2)15-19(21)23/h3-4,8-11,15-16H,5-7,12-14H2,1-2H3 |
| SMILES | CSc1ccc2c(c1)N(CCC3CCCCN3C)c4ccccc4S2=O |
| IUPAC Name | 10-[2-(1-methylpiperidin-2-yl)ethyl]-2-methylsulfanylphenothiazine 5-oxide |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction