missing translation for 'onlineSavingsMsg'
Learn More

Oxcarbazepine EP Impurity C (Oxcarbazepine USP Related Compound E), Mikromol™

Product Code. 30050641
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
This item is not returnable. View return policy

Product Code. 30050641

Brand: Mikromol MM0076.14

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Oxcarbazepine EP Impurity C (Oxcarbazepine USP Related Compound E)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 10-Oxo-10,11-Dihydro-5H-dibenz[b,f]azepine
CAS 21737-58-6
Analyte or Component Names 10-Oxo-10,11-Dihydro-5H-dibenz[b,f]azepine
Shipping Condition Room Temperature
Molecular Formula C14 H11 N O
Certifications/Compliance ISO 17025
Synonym 10-Ketoiminodibenzyl (10-Oxo-10,11-dihydro-5H-dibenz[b,f]azepine), Oxcarbazepine Imp. C (EP), Oxcarbazepine USP Related Compound E, Oxcarbazepine USP RC E, 5,11-Dihydro-10H-dibenz[b,f]azepin-10-one, Oxcarbazepine Related Compound E, Oxcarbazepine Impurity C
InChI Formula InChI=1S/C14H11NO/c16-14-9-10-5-1-3-7-12(10)15-13-8-4-2-6-11(13)14/h1-8,15H,9H2
SMILES O=C1Cc2ccccc2Nc3ccccc13
IUPAC Name 6,11-dihydrobenzo[b][1]benzazepin-5-one
Molecular Weight (g/mol) 209.24
Quantity 100 mg
Formula Weight 209.0841 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.