missing translation for 'onlineSavingsMsg'
Learn More
Learn More
11beta,17,21-Trihydroxypregna-4,6-diene-3,20-dione (Δ6-Hydrocortisone), Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
11beta,17,21-Trihydroxypregna-4,6-diene-3,20-dione (Δ6-Hydrocortisone)
Specifications
Specifications
| Chemical Name or Material | 11beta,17,21-Trihydroxypregna-4,6-diene-3,20-dione (Δ6-Hydrocortisone) |
| CAS | 600-99-7 |
| Analyte or Component Names | 11beta,17,21-Trihydroxypregna-4,6-diene-3,20-dione (Δ6-Hydrocortisone) |
| Shipping Condition | Room Temperature |
| Molecular Formula | C21 H28 O5 |
| Certifications/Compliance | ISO 9001 |
| Synonym | 1-[[4-[2-[(3,5-Di-chloro-2-methoxybenzoyl)amino]ethyl]phenyl]sulfonyl]-3-cyclohexylurea, Glibenclamide Imp. E (Pharmeuropa), Hydrocortisone Imp. E (EP), 11beta,17,21-Trihydroxypregna-4,6-diene-3,20-dione, Delta6-Hydrocortisone, Pregna-4,6-diene-3,20-dione, 11,17,21-trihydroxy-, (11beta)- (9CI), Pregna-4,6-diene-3,20-dione, 11beta,17,21-trihydroxy- (8CI), 6-Dehydrocortisol, NSC 12881, Delta6-Cortisol, Hydrocortisone Impurity E |
| InChI Formula | InChI=1S/C21H28O5/c1-19-7-5-13(23)9-12(19)3-4-14-15-6-8-21(26,17(25)11-22)20(15,2)10-16(24)18(14)19/h3-4,9,14-16,18,22,24,26H,5-8,10-11H2,1-2H3/t14-,15-,16-,18+,19-,20-,21-/m0/s1 |
| SMILES | C[C@]12CCC(=O)C=C1C=C[C@H]3[C@@H]4CC[C@](O)(C(=O)CO)[C@@]4(C)C[C@H](O)[C@H]23 |
| IUPAC Name | (8S,9S,10R,11S,13S,14S,17R)-11,17-dihydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-2,8,9,11,12,14,15,16-octahydro-1H-cyclopenta[a]phenanthren-3-one |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction