missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(1R)-1-Phenyl-1,2,3,4-tetrahydroisoquinoline, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
Description
(1R)-1-Phenyl-1,2,3,4-tetrahydroisoquinoline
Specifications
Specifications
| Chemical Name or Material | (1R)-1-Phenyl-1,2,3,4-tetrahydroisoquinoline |
| CAS | 180272-45-1 |
| Analyte or Component Names | (1R)-1-Phenyl-1,2,3,4-tetrahydroisoquinoline |
| Shipping Condition | Room Temperature |
| Molecular Formula | C15 H15 N |
| Certifications/Compliance | ISO 17025 |
| Synonym | Isoquinoline, 1,2,3,4-tetrahydro-1-phenyl-, (1R)-, Isoquinoline, 1,2,3,4-tetrahydro-1-phenyl-, (R)-, (-)-1-Phenyl-1,2,3,4-tetrahydroisoquinoline, (1R)-1-Phenyl-1,2,3,4-tetrahydroisoquinoline, (1R)-Phenyl-1,2,3,4-tetrahydroisoquinoline, (R)-1-Phenyl-1,2,3,4-tetrahydroisoquinoline |
| InChI Formula | InChI=1S/C15H15N/c1-2-7-13(8-3-1)15-14-9-5-4-6-12(14)10-11-16-15/h1-9,15-16H,10-11H2/t15-/m1/s1 |
| SMILES | C1Cc2ccccc2[C@H](N1)c3ccccc3 |
| IUPAC Name | (1R)-1-phenyl-1,2,3,4-tetrahydroisoquinoline |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction