Learn More
Dipyridamole EP Impurity C, Mikromol™
Product Code. 30050139
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Product Code. 30050139
Brand: Mikromol MM0462.03
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Dipyridamole EP Impurity C
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2,2'-((6-Chloro-4,8-di(piperidin-1-yl)pyrimido[5,4-d]pyrimidin-2-yl)nitrilo)diethanol
CAS 54093-92-4
Analyte or Component Names 2,2'-((6-Chloro-4,8-di(piperidin-1-yl)pyrimido[5,4-d]pyrimidin-2-yl)nitrilo)diethanol
Shipping Condition Room Temperature
Molecular Formula C20 H30 Cl N7 O2
Certifications/Compliance ISO 17025
Synonym 2,2'-[[6-Chloro-4,8-di(piperidin-1-yl)pyrimido[5,4-d]pyrimidin-2-yl]nitrilo]diethanol, Dipyridamole Imp. C (EP), Dipyridamole USP Related Compound C, Dipyridamole USP RC C, 2,2'-[[6-Chloro-4,8-di(piperidin-1-yl)pyrimido[5,4-d]pyrimidin-2-yl]nitrilo]diethanol, Dipyridamole Related Compound C, Dipyridamole Impurity C
InChI Formula InChI=1S/C20H30ClN7O2/c21-19-22-15-16(17(24-19)26-7-3-1-4-8-26)23-20(28(11-13-29)12-14-30)25-18(15)27-9-5-2-6-10-27/h29-30H,1-14H2
SMILES OCCN(CCO)c1nc(N2CCCCC2)c3nc(Cl)nc(N4CCCCC4)c3n1
IUPAC Name 2-[[2-chloro-4,8-di(piperidin-1-yl)pyrimido[5,4-d]pyrimidin-6-yl]-(2-hydroxyethyl)amino]ethanol
Molecular Weight (g/mol) 435.95
Quantity 100 mg
Formula Weight 435.215 g/mol