missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lidocaine EP Impurity E (as Hydrochloride), Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
Lidocaine EP Impurity E (as Hydrochloride)
Specifications
Specifications
| Chemical Name or Material | 2,2'-(Azanediyl)bis[N-(2,6-dimethylphenyl)acetamide] Hydrochloride |
| CAS | 1135231-62-7 |
| Analyte or Component Names | 2,2'-(Azanediyl)bis[N-(2,6-dimethylphenyl)acetamide] Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C20 H25 N3 O2 . Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | Lidocaine Hydrochloride Imp. E (EP), Lidocaine Imp. E (EP), 2,2'-(Azanediyl)bis[N-(2,6-dimethylphenyl)acetamide] Hydrochloride, Lidocaine Hydrochloride Imp. E (EP) as Hydrochloride, Lidocaine Hydrochloride Impurity E as Hydrochloride, Lidocaine Impurity E as Hydrochloride |
| InChI Formula | InChI=1S/C20H25N3O2.ClH/c1-13-7-5-8-14(2)19(13)22-17(24)11-21-12-18(25)23-20-15(3)9-6-10-16(20)4;/h5-10,21H,11-12H2,1-4H3,(H,22,24)(H,23,25);1H |
| SMILES | Cl.Cc1cccc(C)c1NC(=O)CNCC(=O)Nc2c(C)cccc2C |
| IUPAC Name | 2-[[2-(2,6-dimethylanilino)-2-oxoethyl]amino]-N-(2,6-dimethylphenyl)acetamide;hydrochloride |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction