missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(2H-Benzotriazol-2-yl)-4-(1,1,3,3-tetramethylbutyl)phenol, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
2-(2H-Benzotriazol-2-yl)-4-(1,1,3,3-tetramethylbutyl)phenol
Specifications
Specifications
| Chemical Name or Material | 2-(2H-Benzotriazol-2-yl)-4-(1,1,3,3-tetramethylbutyl)phenol |
| CAS | 3147-75-9 |
| Analyte or Component Names | 2-(2H-Benzotriazol-2-yl)-4-(1,1,3,3-tetramethylbutyl)phenol |
| Shipping Condition | Room Temperature |
| Molecular Formula | C20 H25 N3 O |
| Certifications/Compliance | ISO 17025 |
| Synonym | Phenol, 2-(2H-benzotriazol-2-yl)-4-(1,1,3,3-tetramethylbutyl)-, 1268A, 2-(2-Hydroxy-5-t-octylphenyl)-2H-benzotriazole, 2-(2-Hydroxy-5-t-octylphenyl)benzotriazole, 2-(2-Hydroxy-5-tert-octylphenyl)-2H-benzotriazole, 2-(2-Hydroxy-5-tert-octylphenyl)benzotriazole, 2-(2H-Benzotriazol-2-yl)-4-(1,1,3,3-tetramethylbutyl)phenol, 2-(2H-Benzotriazol-2-yl)-4-(2,4,4-trimethylpentan-2-yl)phenol, 2-(2H-Benzotriazol-2-yl)-4-tert-octylphenol, 2-(2H-Benzotriazole-2-yl)-4-(1,1,3,3-tetramethylbutyl)phenol, 2-(2'-Hydroxy-5-tert-octylphenyl)benzotriazole, 2-(2'-Hydroxy-5'-tert-octylphenyl)benzotriazole, 2-(2'-Hydroxy-5'-tert-octylphenyl)benztriazole, 2-(5-t-Octyl-2-hydroxyphenyl)benzotriazole, 2-(5'-tert-Octyl-2'-hydroxyphenyl)benzotriazole, 2-Benzotriazolyl-4-tert-octylphenol, 2-[2'-Hydroxy-5'-(1,1,3,3-tetramethylbutyl)phenyl]benzotriazole, Chisorb 5411, Cyasorb 5411, Cyasorb UV 5411, Eversorb 72, JF 83, Kemisorb 79, Octrizole, Seesorb 709, Seesorb 709G, Spectra-Sorb UV 5411, Sumisorb 340, Tinuvin 329, UV 329, UV 5411, Uvinul 3029, Viosorb 583, Bisoctrizole Related Compound A |
| InChI Formula | InChI=1S/C20H25N3O/c1-19(2,3)13-20(4,5)14-10-11-18(24)17(12-14)23-21-15-8-6-7-9-16(15)22-23/h6-12,24H,13H2,1-5H3 |
| SMILES | CC(C)(C)CC(C)(C)c1ccc(O)c(c1)n2nc3ccccc3n2 |
| IUPAC Name | 2-(benzotriazol-2-yl)-4-(2,4,4-trimethylpentan-2-yl)phenol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction