missing translation for 'onlineSavingsMsg'
Learn More

Doxazosin EP Impurity E, Mikromol™

Product Code. 30052364
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
This item is not returnable. View return policy

Product Code. 30052364

Brand: Mikromol MM0085.06

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Doxazosin EP Impurity E
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,4-Dichloro-6,7-dimethoxyquinazoline
CAS 27631-29-4
Analyte or Component Names 2,4-Dichloro-6,7-dimethoxyquinazoline
Shipping Condition Room Temperature
Molecular Formula C10 H8 Cl2 N2 O2
Certifications/Compliance ISO 17025
Synonym Doxazosin Mesilate Imp. E (EP), Doxazosin USP Related Compound E, Doxazosin USP RC E, 2,4-Dichloro-6,7-dimethoxyquinazoline, Doxazosin Related Compound E, Doxazosin Mesilate Impurity E, Doxazosin Impurity E
InChI Formula InChI=1S/C10H8Cl2N2O2/c1-15-7-3-5-6(4-8(7)16-2)13-10(12)14-9(5)11/h3-4H,1-2H3
SMILES COc1cc2nc(Cl)nc(Cl)c2cc1OC
IUPAC Name 2,4-dichloro-6,7-dimethoxyquinazoline
Molecular Weight (g/mol) 259.09
Quantity 100 mg
Formula Weight 257.9963 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.