missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Benzylprop-2-enoic Acid (2-Benzylacrylic Acid), Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
2-Benzylprop-2-enoic Acid (2-Benzylacrylic Acid)
Specifications
Specifications
| Chemical Name or Material | 2-Benzylacrylic Acid |
| CAS | 5669-19-2 |
| Analyte or Component Names | 2-Benzylacrylic Acid |
| Shipping Condition | Room Temperature |
| Molecular Formula | C10 H10 O2 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Racecadotril Imp. E (EP), Hydrocinnamic acid, alpha-methylene- (6CI,8CI), 2-Benzylacrylic acid, 2-Methylene-3-phenylpropanoic acid, NSC 192640, alpha-Benzylacrylic acid, alpha-Methylenebenzenepropanoic acid, Racecadotril Impurity E |
| InChI Formula | InChI=1S/C10H10O2/c1-8(10(11)12)7-9-5-3-2-4-6-9/h2-6H,1,7H2,(H,11,12) |
| SMILES | OC(=O)C(=C)Cc1ccccc1 |
| IUPAC Name | 2-benzylprop-2-enoic acid |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction