missing translation for 'onlineSavingsMsg'
Learn More
Learn More
[2-Butyl-4-chloro-1-[[2'-[2-(triphenylmethyl)-2H-tetrazol-5-yl]biphenyl-4-yl]methyl]-1H-imidazol-5-yl]methanol, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
Description
[2-Butyl-4-chloro-1-[[2'-[2-(triphenylmethyl)-2H-tetrazol-5-yl]biphenyl-4-yl]methyl]-1H-imidazol-5-yl]methanol
Specifications
Specifications
| Chemical Name or Material | (2-Butyl-4-chloro-1-[(2'-[2-(triphenylmethyl)-2H-tetrazol-5-yl]biphenyl-4-yl)methyl]-1H-imidazol-5-yl)methanol |
| CAS | 133909-99-6 |
| Analyte or Component Names | (2-Butyl-4-chloro-1-[(2'-[2-(triphenylmethyl)-2H-tetrazol-5-yl]biphenyl-4-yl)methyl]-1H-imidazol-5-yl)methanol |
| Shipping Condition | Room Temperature |
| Molecular Formula | C41 H37 Cl N6 O |
| Certifications/Compliance | ISO/IEC 17025 |
| Synonym | Losartan Potassium Imp. H (EP), Losartan Imp. H (EP), [2-Butyl-4-chloro-1-[[2'-[2-(triphenylmethyl)-2H-tetrazol-5-yl]biphenyl-4-yl]methyl]-1H-imidazol-5-yl]methanol, Losartan Potassium Impurity H, Losartan Impurity H |
| InChI Formula | InChI=1S/C41H37ClN6O/c1-2-3-23-38-43-39(42)37(29-49)47(38)28-30-24-26-31(27-25-30)35-21-13-14-22-36(35)40-44-46-48(45-40)41(32-15-7-4-8-16-32,33-17-9-5-10-18-33)34-19-11-6-12-20-34/h4-22,24-27,49H,2-3,23,28-29H2,1H3 |
| SMILES | CCCCc1nc(Cl)c(CO)n1Cc2ccc(cc2)c3ccccc3c4nnn(n4)C(c5ccccc5)(c6ccccc6)c7ccccc7 |
| IUPAC Name | [2-butyl-5-chloro-3-[[4-[2-(2-trityltetrazol-5-yl)phenyl]phenyl]methyl]imidazol-4-yl]methanol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction