missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(2-Chlorophenyl)(1-hydroxycyclopentyl)methanone, Mikromol™
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
(2-Chlorophenyl)(1-hydroxycyclopentyl)methanone
Specifications
Specifications
| Chemical Name or Material | (2-Chlorophenyl)(1-hydroxycyclopentyl)methanone |
| CAS | 90717-17-2 |
| Analyte or Component Names | (2-Chlorophenyl)(1-hydroxycyclopentyl)methanone |
| Shipping Condition | Room Temperature |
| Molecular Formula | C12 H13 Cl O2 |
| Certifications/Compliance | ISO 17025 |
| Synonym | o-Chlorophenyl 1-Hydroxycyclopentyl Ketone, Ketamine Impurity C, Ketamine Hydrochloride Imp. C (EP), Ketamine Imp. C (EP), (2-Chlorophenyl)(1-hydroxycyclopentyl)methanone, 1-Hydroxycyclopentyl 2-chlorophenyl ketone, o-Chlorophenyl 1-hydroxycyclopentyl ketone, Ketamine Hydrochloride Impurity C, Ketamine Impurity C |
| InChI Formula | InChI=1S/C12H13ClO2/c13-10-6-2-1-5-9(10)11(14)12(15)7-3-4-8-12/h1-2,5-6,15H,3-4,7-8H2 |
| SMILES | OC1(CCCC1)C(=O)c2ccccc2Cl |
| IUPAC Name | (2-chlorophenyl)-(1-hydroxycyclopentyl)methanone |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction