missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Flunixin Meglumine EP Impurity B, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
Flunixin Meglumine EP Impurity B
Specifications
Specifications
| Chemical Name or Material | 2-methyl-3-(trifluoromethyl)aniline |
| CAS | 54396-44-0 |
| Analyte or Component Names | 2-methyl-3-(trifluoromethyl)aniline |
| Shipping Condition | Room Temperature |
| Molecular Formula | C8 H8 F3 N |
| Certifications/Compliance | ISO 17025 |
| Synonym | 2-Methyl-3-(trifluoromethyl)aniline, Flunixin Meglumine Imp. B (EP), Flunixin USP Related Compound B, Flunixin USP RC B, 2-Amino-6-(trifluoromethyl)toluene, 2-Methyl-3-(trifluoromethyl)aniline, 2-Methyl-3-(trifluoromethyl)benzenamine, 2-Methyl-3-(trifluoromethyl)phenylamine, 2-Methyl-3-aminobenzotrifluoride, 3-(Trifluoromethyl)-2-methylaniline, 3-(Trifluoromethyl)-2-methylbenzenamine, Flunixin Related Compound B, Flunixin Meglumine Impurity B, Flunixin Impurity B |
| InChI Formula | InChI=1S/C8H8F3N/c1-5-6(8(9,10)11)3-2-4-7(5)12/h2-4H,12H2,1H3 |
| SMILES | Cc1c(N)cccc1C(F)(F)F |
| IUPAC Name | 2-methyl-3-(trifluoromethyl)aniline |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction