missing translation for 'onlineSavingsMsg'
Learn More

Flupentixol EP Impurity G, Mikromol™

Product Code. 17998274
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
This item is not returnable. View return policy

Product Code. 17998274

Brand: Mikromol MM0182.08

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Flupentixol EP Impurity G
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Trifluoromethylthioxanthone
CAS 1693-28-3
Analyte or Component Names 2-Trifluoromethylthioxanthone
Shipping Condition Room Temperature
Molecular Formula C14 H7 F3 O S
Certifications/Compliance ISO 17025
Synonym Flupentixol Dihydrochloride Imp. G (EP), Flupentixol Imp. G (EP), 2-(Trifluoromethyl)-9H-thioxanthen-9-one, 2-Trifluoromethylthioxanthone, Flupentixol Decanoate Imp. B (BP), Flupentixol Dihydrochloride Impurity G, Flupentixol Impurity G
InChI Formula InChI=1S/C14H7F3OS/c15-14(16,17)8-5-6-12-10(7-8)13(18)9-3-1-2-4-11(9)19-12/h1-7H
SMILES FC(F)(F)c1ccc2Sc3ccccc3C(=O)c2c1
IUPAC Name 2-(trifluoromethyl)thioxanthen-9-one
Molecular Weight (g/mol) 280.26
Quantity 100 mg
Formula Weight 280.017 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.