missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(2RS)-1-[4-[(EZ)-2-Methoxyethenyl]-phenoxy]-3-[(1-methylethyl)amino]propan-2-ol, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
(2RS)-1-[4-[(EZ)-2-Methoxyethenyl]-phenoxy]-3-[(1-methylethyl)amino]propan-2-ol
Specifications
Specifications
| Chemical Name or Material | (2RS)-1-[4-[(EZ)-2-Methoxyethenyl]-phenoxy]-3-[(1-methylethyl)amino]propan-2-ol |
| CAS | 82961-02-2 |
| Analyte or Component Names | (2RS)-1-[4-[(EZ)-2-Methoxyethenyl]-phenoxy]-3-[(1-methylethyl)amino]propan-2-ol |
| Shipping Condition | Room Temperature |
| Molecular Formula | C15 H23 N O3 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Tamoxifen Citrate Imp. F (EP), Tamoxifen Imp. F (EP), (2RS)-1-[4-[(EZ)-2-Methoxyethenyl]phenoxy]-3-[(1methylethyl)amino]propan-2-ol |
| InChI Formula | InChI=1S/C15H23NO3/c1-12(2)16-10-14(17)11-19-15-6-4-13(5-7-15)8-9-18-3/h4-9,12,14,16-17H,10-11H2,1-3H3 |
| SMILES | COC=Cc1ccc(OCC(O)CNC(C)C)cc1 |
| IUPAC Name | 1-[4-(2-methoxyethenyl)phenoxy]-3-(propan-2-ylamino)propan-2-ol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction