missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promethazine EP Impurity D, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
Promethazine EP Impurity D
Specifications
Specifications
| Chemical Name or Material | Promethazine Sulfoxide |
| CAS | 7640-51-9 |
| Analyte or Component Names | Promethazine Sulfoxide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C17 H20 N2 O S |
| Certifications/Compliance | ISO 17025 |
| Synonym | (2RS)-N,N-Dimethyl-1-(10H-phenothiazin-10-yl)propan-2-amine S-Oxide (Promethazine Sulphoxide), (2RS)-N,N-Dimethyl-1-(10H-phenothiazin-10-yl)propan-2-amine S-Oxide, Promethazine Hydrochloride Imp. D (EP), Promethazine Sulphoxide, Promethazine Hydrochloride Impurity D, Promethazine Impurity D |
| InChI Formula | InChI=1S/C17H20N2OS/c1-13(18(2)3)12-19-14-8-4-6-10-16(14)21(20)17-11-7-5-9-15(17)19/h4-11,13H,12H2,1-3H3 |
| SMILES | CC(CN1c2ccccc2S(=O)c3ccccc13)N(C)C |
| IUPAC Name | N,N-dimethyl-1-(5-oxophenothiazin-10-yl)propan-2-amine |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction