missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-[1-[4-(Aminosulfonyl)phenyl]-3-(trifluoromethyl)-1H-pyrazol-5-yl]benzoic Acid, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
Description
4-[1-[4-(Aminosulfonyl)phenyl]-3-(trifluoromethyl)-1H-pyrazol-5-yl]benzoic Acid
Specifications
Specifications
| Chemical Name or Material | Celecoxib Carboxylic Acid |
| CAS | 170571-01-4 |
| Analyte or Component Names | Celecoxib Carboxylic Acid |
| Shipping Condition | Room Temperature |
| Molecular Formula | C17 H12 F3 N3 O4 S |
| Certifications/Compliance | ISO 17025 |
| Synonym | 4-[1-[4-(Aminosulfonyl)phenyl]-3-(trifluoromethyl)-1H-pyrazol-5-yl]benzoic Acid, Celecoxib Carboxylic Acid |
| InChI Formula | InChI=1S/C17H12F3N3O4S/c18-17(19,20)15-9-14(10-1-3-11(4-2-10)16(24)25)23(22-15)12-5-7-13(8-6-12)28(21,26)27/h1-9H,(H,24,25)(H2,21,26,27) |
| SMILES | NS(=O)(=O)c1ccc(cc1)n2nc(cc2c3ccc(cc3)C(=O)O)C(F)(F)F |
| IUPAC Name | 4-[2-(4-sulfamoylphenyl)-5-(trifluoromethyl)pyrazol-3-yl]benzoic acid |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction