missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-[4-[4-[4-[[cis-2-(2,4-Dichlorophenyl)-2-(1H-1,2,4-triazol-1-ylmethyl)-1,3-dioxolan-4-yl]methoxy]phenyl]piperazin-1-yl]phenyl]-2-propyl-2,4-dihydro-3H-1,2,4-triazol-3-one, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
Description
4-[4-[4-[4-[[cis-2-(2,4-Dichlorophenyl)-2-(1H-1,2,4-triazol-1-ylmethyl)-1,3-dioxolan-4-yl]methoxy]phenyl]piperazin-1-yl]phenyl]-2-propyl-2,4-dihydro-3H-1,2,4-triazol-3-one
Specifications
Specifications
| Chemical Name or Material | 4-[4-[4-[4-[[cis-2-(2,4-Dichlorophenyl)-2-(1H-1,2,4-triazol-1-ylmethyl)-1,3-dioxolan-4-yl]methoxy]phenyl]piperazin-1-yl]phenyl]-2-propyl-2,4-dihydro-3H-1,2,4-triazol-3-one |
| CAS | 74855-91-7 |
| Analyte or Component Names | 4-[4-[4-[4-[[cis-2-(2,4-Dichlorophenyl)-2-(1H-1,2,4-triazol-1-ylmethyl)-1,3-dioxolan-4-yl]methoxy]phenyl]piperazin-1-yl]phenyl]-2-propyl-2,4-dihydro-3H-1,2,4-triazol-3-one |
| Shipping Condition | Room Temperature |
| Molecular Formula | C34 H36 Cl2 N8 O4 |
| Certifications/Compliance | ISO 9001 |
| Synonym | 4-[4-[4-[4-[[cis-2-(2,4-Dichlorophenyl)-2-(1H-1,2,4-triazol-1-ylmethyl)-1,3-dioxolan-4-yl]methoxy]phenyl]piperazin-1-yl]phenyl]-2-propyl-2,4-dihydro-3H-1,2,4-triazol-3-one, Itraconazole Imp. C (EP), Itraconazole Impurity C |
| InChI Formula | InChI=1S/C34H36Cl2N8O4/c1-2-13-44-33(45)43(24-39-44)28-6-4-26(5-7-28)40-14-16-41(17-15-40)27-8-10-29(11-9-27)46-19-30-20-47-34(48-30,21-42-23-37-22-38-42)31-12-3-25(35)18-32(31)36/h3-12,18,22-24,30H,2,13-17,19-21H2,1H3/t30-,34-/m0/s1 |
| SMILES | CCCN1N=CN(C1=O)c2ccc(cc2)N3CCN(CC3)c4ccc(OC[C@H]5CO[C@@](Cn6cncn6)(O5)c7ccc(Cl)cc7Cl)cc4 |
| IUPAC Name | 4-[4-[4-[4-[[(2R,4S)-2-(2,4-dichlorophenyl)-2-(1,2,4-triazol-1-ylmethyl)-1,3-dioxolan-4-yl]methoxy]phenyl]piperazin-1-yl]phenyl]-2-propyl-1,2,4-triazol-3-one |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction