missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Acetylaminophenazone, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
4-Acetylaminophenazone
Specifications
Specifications
| Chemical Name or Material | 4-Acetylaminophenazone |
| CAS | 83-15-8 |
| Analyte or Component Names | 4-Acetylaminophenazone |
| Shipping Condition | Room Temperature |
| Molecular Formula | C13 H15 N3 O2 |
| Certifications/Compliance | ISO 17025 |
| Synonym | 4-Acetylaminophenazone, Acetamide, N-(2,3-dihydro-1,5-dimethyl-3-oxo-2-phenyl-1H-pyrazol-4-yl)- (9CI), Acetamide, N-antipyrinyl- (8CI), Antipyrine, 4-acetamido- (6CI,7CI), 4-(N-Acetylamino)antipyrine, 4-Acetamido-2,3-dimethyl-1-phenylpyrazol-5-one, 4-Acetamidoantipyrine, 4-Acetaminoantipyrine, 4-Acetoaminoantipyrine, 4-Acetylaminoantipyrine, Acetylaminoantipyrine, N-Acetyl-4-aminoantipyrine, N-Acetyl-4-aminophenazone, N-Antipyrinylacetamide, NSC 331807 |
| InChI Formula | InChI=1S/C13H15N3O2/c1-9-12(14-10(2)17)13(18)16(15(9)3)11-7-5-4-6-8-11/h4-8H,1-3H3,(H,14,17) |
| SMILES | CN1N(C(=O)C(=C1C)NC(=O)C)c2ccccc2 |
| IUPAC Name | N-(1,5-dimethyl-3-oxo-2-phenylpyrazol-4-yl)acetamide |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction