missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Amino-5-hydroxymethyl-2-methylpyrimidine Hydrochloride, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
4-Amino-5-hydroxymethyl-2-methylpyrimidine Hydrochloride
Specifications
Specifications
| Chemical Name or Material | 4-Amino-5-hydroxymethyl-2-methylpyrimidine Hydrochloride |
| CAS | 13100-57-7 |
| Analyte or Component Names | 4-Amino-5-hydroxymethyl-2-methylpyrimidine Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C6 H9 N3 O . Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | 4-Amino-5-hydroxymethyl-2-methylpyrimidine Hydrochloride |
| InChI Formula | InChI=1S/C6H9N3O.ClH/c1-4-8-2-5(3-10)6(7)9-4;/h2,10H,3H2,1H3,(H2,7,8,9);1H |
| SMILES | Cl.Cc1ncc(CO)c(N)n1 |
| IUPAC Name | (4-amino-2-methylpyrimidin-5-yl)methanol;hydrochloride |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction