missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-1,3,4-thiadiazol-2-thiol (MMTD), Mikromol™

Product Code. 17924186
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
This item is not returnable. View return policy

Product Code. 17924186

Brand: Mikromol MM0238.05

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

5-Methyl-1,3,4-thiadiazol-2-thiol (MMTD)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-Methyl-1,3,4-thiadiazol-2-thiol (MMTD)
CAS 29490-19-5
Analyte or Component Names 5-Methyl-1,3,4-thiadiazol-2-thiol (MMTD)
Shipping Condition Room Temperature
Molecular Formula C3 H4 N2 S2
Certifications/Compliance ISO 17025
Synonym Cefazolin Sodium Imp. E (EP), Cefazolin Imp. E (EP), 5-Methyl-1,3,4-thiadiazol-2-thiol, MMTD, Cefazolin Sodium Impurity E, Cefazolin Impurity E
InChI Formula InChI=1S/C3H4N2S2/c1-2-4-5-3(6)7-2/h1H3,(H,5,6)
SMILES Cc1nnc(S)s1
IUPAC Name 5-methyl-1,3,4-thiadiazole-2-thiol
Molecular Weight (g/mol) 132.21
Quantity 100 mg
Formula Weight 131.9816 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.