missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(5R)-3-(3-Fluoro-4-morpholinophenyl)-5-(hydroxymethyl)oxazolidin-2-one, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
(5R)-3-(3-Fluoro-4-morpholinophenyl)-5-(hydroxymethyl)oxazolidin-2-one
Specifications
Specifications
| Chemical Name or Material | (5R)-3-(3-Fluoro-4-morpholinophenyl)-5-(hydroxymethyl)oxazolidin-2-one |
| CAS | 168828-82-8 |
| Analyte or Component Names | (5R)-3-(3-Fluoro-4-morpholinophenyl)-5-(hydroxymethyl)oxazolidin-2-one |
| Shipping Condition | Room Temperature |
| Molecular Formula | C14 H17 F N2 O4 |
| Certifications/Compliance | ISO 17025 |
| Synonym | 2-Oxazolidinone, 3-[3-fluoro-4-(4-morpholinyl)phenyl]-5-(hydroxymethyl)-, (5R)-, 2-Oxazolidinone, 3-[3-fluoro-4-(4-morpholinyl)phenyl]-5-(hydroxymethyl)-, (R)-, (5R)-5-(Hydroxymethyl)-3-[3-fluoro-4-(4-morpholinyl)phenyl]-2-oxazolidinone, (R)-5-(Hydroxymethyl)-3-(3-fluoro-4-morpholinophenyl)oxazolidin-2-one, (R)-[3-(3-Fluoro-4-morpholin-4-ylphenyl)-2-oxo-5-oxazolidinyl]methanol, (R)-[3-(3-Fluoro-4-morpholinophenyl)-2-oxo-5-oxazolidinyl]methanol, [(R)-3-(3-Fluoro-4-morpholinophenyl)-2-oxo-5-oxazolidinyl]methanol |
| InChI Formula | InChI=1S/C14H17FN2O4/c15-12-7-10(17-8-11(9-18)21-14(17)19)1-2-13(12)16-3-5-20-6-4-16/h1-2,7,11,18H,3-6,8-9H2/t11-/m1/s1 |
| SMILES | OC[C@H]1CN(C(=O)O1)c2ccc(N3CCOCC3)c(F)c2 |
| IUPAC Name | (5R)-3-(3-fluoro-4-morpholin-4-ylphenyl)-5-(hydroxymethyl)-1,3-oxazolidin-2-one |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction