missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Chloro-4-phenyl-3,4-dihydroquinazoline-2-carboxylic Acid, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
6-Chloro-4-phenyl-3,4-dihydroquinazoline-2-carboxylic Acid
Specifications
Specifications
| Chemical Name or Material | 6-Chloro-4-phenyl-3,4-dihydroquinazoline-2-carboxylic Acid |
| CAS | 88190-77-6 |
| Analyte or Component Names | 6-Chloro-4-phenyl-3,4-dihydroquinazoline-2-carboxylic Acid |
| Shipping Condition | Room Temperature |
| Molecular Formula | C15 H11 Cl N2 O2 |
| Certifications/Compliance | ISO 9001 |
| Synonym | 6-Chloro-4-phenyl-3,4-dihydroquinazoline-2-carboxylic Acid |
| InChI Formula | InChI=1S/C15H11ClN2O2/c16-10-6-7-12-11(8-10)13(9-4-2-1-3-5-9)18-14(17-12)15(19)20/h1-8,13H,(H,17,18)(H,19,20) |
| SMILES | OC(=O)C1=Nc2ccc(Cl)cc2C(N1)c3ccccc3 |
| IUPAC Name | 6-chloro-4-phenyl-3,4-dihydroquinazoline-2-carboxylic acid |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction