missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6beta-Hydroxy-6-methyl-3,20-dioxopregn-4-en-17-yl Acetate (6beta-Hydroxymedroxyprogesterone Acetate), Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
Description
6beta-Hydroxy-6-methyl-3,20-dioxopregn-4-en-17-yl Acetate (6beta-Hydroxymedroxyprogesterone Acetate)
Specifications
Specifications
| Chemical Name or Material | 6beta-Hydroxymedroxyprogesterone Acetate |
| CAS | 984-47-4 |
| Analyte or Component Names | 6beta-Hydroxymedroxyprogesterone Acetate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C24 H34 O5 |
| Certifications/Compliance | ISO 17025 |
| Synonym | 6beta-Hydroxy-6-methyl-3,20-dioxopregn-4-en-17-yl Acetate, Medroxyprogesterone Acetate Imp. A (EP), 6beta-Hydroxymedroxyprogesterone Acetate, Pregn-4-ene-3,20-dione, 17-(acetyloxy)-6-hydroxy-6-methyl-, (6beta)-, Pregn-4-ene-3,20-dione, 6beta,17-dihydroxy-6-methyl-, 17-acetate (7CI,8CI), (6beta)-17-(Acetyloxy)-6-hydroxy-6-methylpregn-4-ene-3,20-dione, Medroxyprogesterone Acetate Impurity A |
| InChI Formula | InChI=1S/C24H34O5/c1-14(25)24(29-15(2)26)11-8-19-17-13-23(5,28)20-12-16(27)6-9-21(20,3)18(17)7-10-22(19,24)4/h12,17-19,28H,6-11,13H2,1-5H3/t17-,18+,19+,21-,22+,23-,24+/m1/s1 |
| SMILES | CC(=O)O[C@@]1(CC[C@H]2[C@@H]3C[C@@](C)(O)C4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C)C(=O)C |
| IUPAC Name | [(6R,8R,9S,10R,13S,14S,17R)-17-acetyl-6-hydroxy-6,10,13-trimethyl-3-oxo-1,2,7,8,9,11,12,14,15,16-decahydrocyclopenta[a]phenanthren-17-yl] acetate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction