missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9,11beta-Epoxy-17,21-dihydroxy-16beta-methyl-9beta-pregna-1,4-diene-3,20-dione, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
9,11beta-Epoxy-17,21-dihydroxy-16beta-methyl-9beta-pregna-1,4-diene-3,20-dione
Specifications
Specifications
| Chemical Name or Material | 9,11beta-Epoxy-17,21-dihydroxy-16beta-methyl-9beta-pregna-1,4-diene-3,20-dione |
| CAS | 981-34-0 |
| Analyte or Component Names | 9,11beta-Epoxy-17,21-dihydroxy-16beta-methyl-9beta-pregna-1,4-diene-3,20-dione |
| Shipping Condition | Room Temperature |
| Molecular Formula | C22 H28 O5 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Betamethasone Imp. E (EP), Pregna-1,4-diene-3,20-dione, 9,11-epoxy-17,21-dihydroxy-16-methyl-, (9beta,11beta,16beta)-, 9beta-Pregna-1,4-diene-3,20-dione, 9,11beta-epoxy-17,21-dihydroxy-16beta-methyl- (6CI,7CI,8CI), 9,11-Epoxy-9H-cyclopenta[a]phenanthrene, pregna-1,4-diene-3,20-dione deriv., 9,11beta-Epoxy-17,21-dihydroxy-16beta-methyl-9beta-pregna-1,4-diene-3,20-dione, Betamethasone 9,11-epoxide, Beclometasone Dipropionate Impurity R, Betamethasone Impurity E |
| InChI Formula | InChI=1S/C22H28O5/c1-12-8-16-15-5-4-13-9-14(24)6-7-19(13,2)22(15)18(27-22)10-20(16,3)21(12,26)17(25)11-23/h6-7,9,12,15-16,18,23,26H,4-5,8,10-11H2,1-3H3/t12-,15-,16-,18-,19-,20-,21-,22+/m0/s1 |
| SMILES | C[C@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]35O[C@H]5C[C@]2(C)[C@@]1(O)C(=O)CO |
| IUPAC Name | 9,11β-Epoxy-17,21-dihydroxy-16β-methyl-9β-pregna-1,4-diene-3,20-dione |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction