Learn More
Acrylamide-d3, TRC
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Chemical Name or Material | Acrylamide-d3 |
| CAS | 122775-19-3 |
| Molecular Formula | C3 2H3 H2 N O |
| Quantity | 100 mg |
| Synonym | 2-Propenamide-2,3,3-d3, Acrylamide-2,3,3 D3, Acrylamide-d3, Acrylamide-2,2,2-D3 |
| InChI Formula | InChI=1S/C3H5NO/c1-2-3(4)5/h2H,1H2,(H2,4,5)/i1D2,2D |
| SMILES | [2H]C(=C([2H])C(=O)N)[2H] |
| IUPAC Name | 2,3,3-trideuterioprop-2-enamide |
| Molecular Weight (g/mol) | 74.1 |
| Formula Weight | 74.0559 |