missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-Heptadecafluorooctane-1-sulfonate (~80%), TRC
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Chemical Name or Material | Ammonium 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-Heptadecafluorooctane-1-sulfonate |
| CAS | 29081-56-9 |
| Molecular Formula | C8H4F17NO3S |
| Quantity | 100 mg |
| Synonym | 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluoro-1-Octanesulfonic acid ammonium salt (1:1), 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluoro-1-Octanesulfonic acid ammonium salt (8CI,9CI), Ammonium perfluorooctylsulfonate, EF 104, Eftop EF 104, FC 93, Fluorad FC 93, Perfluorooctanesulfonic acid, ammonium salt |
| InChI Formula | InChI=1S/C8HF17O3S.H3N/c9-1(10,3(13,14)5(17,18)7(21,22)23)2(11,12)4(15,16)6(19,20)8(24,25)29(26,27)28;/h(H,26,27,28);1H3 |
| SMILES | N.O[S](=O)(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F |
| IUPAC Name | azanium;1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluorooctane-1-sulfonate |
| Molecular Weight (g/mol) | 517.16 |
| Formula Weight | 516.964 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction