Learn More
Ampeloptin, TRC
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Chemical Name or Material | Dihydromyricetin |
| CAS | 27200-12-0 |
| Molecular Formula | C15 H12 O8 |
| Quantity | 100 mg |
| Synonym | 4H-1-Benzopyran-4-one, 2,3-dihydro-3,5,7-trihydroxy-2-(3,4,5-trihydroxyphenyl)-, (2R,3R)-, 4H-1-Benzopyran-4-one, 2,3-dihydro-3,5,7-trihydroxy-2-(3,4,5-trihydroxyphenyl)-, (2R-trans)-, Ampelopsin (6CI), Flavanone, 3,3',4',5,5',7-hexahydroxy- (7CI,8CI), (+)-Ampelopsin, (+)-Dihydromyricetin, (2R,3R)-2,3-Dihydro-3,5,7-trihydroxy-2-(3,4,5-trihydroxyphenyl)-4H-1-benzopyran-4-one, Ampelopsin (flavanol), Ampeloptin, Dihydromyricetin |
| InChI Formula | InChI=1S/C15H12O8/c16-6-3-7(17)11-10(4-6)23-15(14(22)13(11)21)5-1-8(18)12(20)9(19)2-5/h1-4,14-20,22H/t14-,15+/m0/s1 |
| SMILES | O[C@@H]1[C@H](Oc2cc(O)cc(O)c2C1=O)c3cc(O)c(O)c(O)c3 |
| IUPAC Name | (2R,3R)-3,5,7-trihydroxy-2-(3,4,5-trihydroxyphenyl)-2,3-dihydrochromen-4-one |
| Molecular Weight (g/mol) | 320.25 |
| Formula Weight | 320.0532 |