missing translation for 'onlineSavingsMsg'
Learn More

Androsta-5,16-diene-3beta,17-diol 3-Acetate 17-Trifluoromethanesulfonate, Mikromol™

Product Code. 30051891
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
This item is not returnable. View return policy

Product Code. 30051891

Brand: Mikromol MM3504.04

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Androsta-5,16-diene-3beta,17-diol 3-Acetate 17-Trifluoromethanesulfonate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Androsta-5,16-diene-3beta,17-diol 3-Acetate 17-Trifluoromethanesulfonate
CAS 115375-60-5
Analyte or Component Names Androsta-5,16-diene-3beta,17-diol 3-Acetate 17-Trifluoromethanesulfonate
Shipping Condition Room Temperature
Molecular Formula C22 H29 F3 O5 S
Certifications/Compliance ISO 17025
Synonym 3β-Acetoxyandrosta-5,16-dien-17-yl trifluoromethanesulfonate, Androsta-5,16-diene-3,17-diol, 3-acetate 17-(trifluoromethanesulfonate), (3β)-, (3β)-Acetoxy-17-(3-trifluoromethylsulfonyloxy)androsta-5,16 diene
InChI Formula InChI=1S/C22H29F3O5S/c1-13(26)29-15-8-10-20(2)14(12-15)4-5-16-17-6-7-19(21(17,3)11-9-18(16)20)30-31(27,28)22(23,24)25/h4,7,15-18H,5-6,8-12H2,1-3H3/t15-,16-,17-,18-,20-,21-/m0/s1
SMILES CC(=O)O[C@H]1CC[C@]2(C)[C@H]3CC[C@@]4(C)[C@@H](CC=C4OS(=O)(=O)C(F)(F)F)[C@@H]3CC=C2C1
IUPAC Name [(3S,8R,9S,10R,13S,14S)-10,13-dimethyl-17-(trifluoromethylsulfonyloxy)-2,3,4,7,8,9,11,12,14,15-decahydro-1H-cyclopenta[a]phenanthren-3-yl] acetate
Molecular Weight (g/mol) 462.52
Quantity 100 mg
Formula Weight 462.1688 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.