missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Flunarizine N-Oxide Dihydrochloride, Mikromol™
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
Flunarizine N-Oxide Dihydrochloride
Specifications
Specifications
| Chemical Name or Material | Flunarizine N-Oxide Dihydrochloride |
| Analyte or Component Names | Flunarizine N-Oxide Dihydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C26 H26 F2 N2 O . 2 Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | Flunarizine N-Oxide Dihydrochloride |
| InChI Formula | InChI=1S/C26H26F2N2O.2ClH/c27-24-12-8-22(9-13-24)26(23-10-14-25(28)15-11-23)29-16-19-30(31,20-17-29)18-4-7-21-5-2-1-3-6-21;;/h1-15,26H,16-20H2;2*1H/b7-4+;; |
| SMILES | Cl.Cl.[O-][N+]1(C\C=C\c2ccccc2)CCN(CC1)C(c3ccc(F)cc3)c4ccc(F)cc4 |
| IUPAC Name | 4-[bis(4-fluorophenyl)methyl]-1-oxido-1-[(E)-3-phenylprop-2-enyl]piperazin-1-ium;dihydrochloride |
| Molecular Weight (g/mol) | 493.42 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction