missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isopropyl 4-Acetylbenzoate, Mikromol™
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
Isopropyl 4-Acetylbenzoate
Specifications
Specifications
| Chemical Name or Material | Isopropyl 4-Acetylbenzoate |
| CAS | 220089-22-5 |
| Analyte or Component Names | Isopropyl 4-Acetylbenzoate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C12 H14 O3 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Benzoic acid, 4-acetyl-, 1-methylethyl ester, 1-Methylethyl 4-acetylbenzoate |
| InChI Formula | InChI=1S/C12H14O3/c1-8(2)15-12(14)11-6-4-10(5-7-11)9(3)13/h4-8H,1-3H3 |
| SMILES | CC(C)OC(=O)c1ccc(cc1)C(=O)C |
| IUPAC Name | propan-2-yl 4-acetylbenzoate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction