missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Acetyl-L-Thyroxine (N-Acetyllevothyroxine), Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
N-Acetyl-L-Thyroxine (N-Acetyllevothyroxine)
Specifications
Specifications
| Chemical Name or Material | N-Acetyl-L-Thyroxine |
| CAS | 26041-51-0 |
| Analyte or Component Names | N-Acetyl-L-Thyroxine |
| Shipping Condition | Room Temperature |
| Molecular Formula | C17 H13 I4 N O5 |
| Certifications/Compliance | ISO 9001 |
| Synonym | L-Tyrosine, N-acetyl-O-(4-hydroxy-3,5-diiodophenyl)-3,5-diiodo-, Thyroxine, N-acetyl- (6CI,7CI), Thyroxine, N-acetyl-, L- (8CI), N-Acetyl-L-thyroxine, N-Acetylthyroxine |
| InChI Formula | InChI=1S/C17H13I4NO5/c1-7(23)22-14(17(25)26)4-8-2-12(20)16(13(21)3-8)27-9-5-10(18)15(24)11(19)6-9/h2-3,5-6,14,24H,4H2,1H3,(H,22,23)(H,25,26)/t14-/m0/s1 |
| SMILES | CC(=O)N[C@@H](Cc1cc(I)c(Oc2cc(I)c(O)c(I)c2)c(I)c1)C(=O)O |
| IUPAC Name | (2S)-2-acetamido-3-[4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl]propanoic acid |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction