missing translation for 'onlineSavingsMsg'
Learn More

N-Cyano-3-[[[2-[(diaminomethylene)amino]thiazol-4-yl]methyl]sulfanyl]propanimidamide Maleate, Mikromol™

Product Code. 30051499
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
This item is not returnable. View return policy

Product Code. 30051499

Brand: Mikromol MM0029.09

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

N-Cyano-3-[[[2-[(diaminomethylene)amino]thiazol-4-yl]methyl]sulfanyl]propanimidamide Maleate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Cyano-3-[((2-[(diaminomethylene)amino]thiazol-4-yl)methyl)sulfanyl]propanimidamide Maleate
Analyte or Component Names N-Cyano-3-[((2-[(diaminomethylene)amino]thiazol-4-yl)methyl)sulfanyl]propanimidamide Maleate
Shipping Condition Room Temperature
Molecular Formula C9 H13 N7 S2 . C4 H4 O4
Certifications/Compliance ISO 17025
Synonym Famotidine Imp. G (EP), N-Cyano-3-[[[2-(diaminomethylene)amino]thiazol-4-yl]methyl]sulfanyl]propanimidamide Maleate, Famotidine Imp. G (EP) as Maleate, Famotidine Impurity G as Maleate
InChI Formula InChI=1S/C9H13N7S2.C4H4O4/c10-5-14-7(11)1-2-17-3-6-4-18-9(15-6)16-8(12)13;5-3(6)1-2-4(7)8/h4H,1-3H2,(H2,11,14)(H4,12,13,15,16);1-2H,(H,5,6)(H,7,8)/b;2-1-
SMILES NC(=Nc1nc(CSCCC(=N)NC#N)cs1)N.OC(=O)\C=C/C(=O)O
IUPAC Name (Z)-but-2-enedioic acid;N-cyano-3-[[2-(diaminomethylideneamino)-1,3-thiazol-4-yl]methylsulfanyl]propanimidamide
Molecular Weight (g/mol) 399.45
Quantity 100 mg
Formula Weight 399.0783 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.