missing translation for 'onlineSavingsMsg'
Learn More

N-Desmethylolanzapine, Mikromol™

Product Code. 17943436
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
This item is not returnable. View return policy

Product Code. 17943436

Brand: Mikromol MM0329.01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

N-Desmethylolanzapine
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Desmethylolanzapine
CAS 161696-76-0
Analyte or Component Names Desmethylolanzapine
Shipping Condition Room Temperature
Molecular Formula C16 H18 N4 S
Certifications/Compliance ISO 17025
Synonym N-Desmethylolanzapine, Desmethylolanzapine, 2-Methyl-4-(1-piperazinyl)-10H-thieno[2,3-b][1,5]benzodiazepine, 4'-Desmethylolanzapine, LY 170055, N-Demethylolanzapine
InChI Formula InChI=1S/C16H18N4S/c1-11-10-12-15(20-8-6-17-7-9-20)18-13-4-2-3-5-14(13)19-16(12)21-11/h2-5,10,17,19H,6-9H2,1H3
SMILES Cc1cc2C(=Nc3ccccc3Nc2s1)N4CCNCC4
IUPAC Name 2-methyl-4-piperazin-1-yl-10H-thieno[2,3-b][1,5]benzodiazepine
Molecular Weight (g/mol) 298.41
Quantity 100 mg
Formula Weight 298.1252 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.