missing translation for 'onlineSavingsMsg'
Learn More

Pralidoxime Methyl Sulphate, Mikromol™

Product Code. 17980966
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 17980966

Brand: Mikromol MM0689.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Pralidoxime Methyl Sulphate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Pralidoxime Methyl Sulphate
CAS 1200-55-1
Analyte or Component Names Pralidoxime Methyl Sulphate
Shipping Condition Ice pack (-20°C)
Molecular Formula C7 H9 N2 O . C H3 O4 S
Certifications/Compliance ISO 17034
Synonym Pralidoxime Methyl Sulphate
InChI Formula InChI=1S/C7H8N2O.CH4O4S/c1-9-5-3-2-4-7(9)6-8-10;1-5-6(2,3)4/h2-6H,1H3;1H3,(H,2,3,4)
SMILES COS(=O)(=O)[O-].C[n+]1ccccc1\C=N\O
IUPAC Name (NE)-N-[(1-methylpyridin-1-ium-2-yl)methylidene]hydroxylamine;methyl sulfate
Molecular Weight (g/mol) 248.26
Quantity 100 mg
Formula Weight 248.0467 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.