missing translation for 'onlineSavingsMsg'
Learn More

Pteroic Acid (>85%), TRC

Product Code. 30005059
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30005059 100 mg 100mg
30135982 50 mg 50mg
30011736 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30005059 Supplier Toronto Research Chemicals Supplier No. TRCP840110100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 119-24-4
Molecular Formula C14 H12 N6 O3
Molecular Weight (g/mol) 312.28
Synonym 4-[[(2-Amino-4-oxo-1,4-dihydropteridin-6-yl)methyl]amino]benzoic Acid, Folic Acid Imp. D (EP), Pteroic Acid
IUPAC Name 4-[(2-amino-4-oxo-1H-pteridin-6-yl)methylamino]benzoic acid
SMILES NC1=NC(=O)c2nc(CNc3ccc(cc3)C(=O)O)cnc2N1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C14H12N6O3/c15-14-19-11-10(12(21)20-14)18-9(6-17-11)5-16-8-3-1-7(2-4-8)13(22)23/h1-4,6,16H,5H2,(H,22,23)(H3,15,17,19,20,21)
Formula Weight 312.0971
Chemical Name or Material Pteroic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.