missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thiazolsulfone, Mikromol™
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
Thiazolsulfone
Specifications
Specifications
| Chemical Name or Material | Thiazolsulfone |
| CAS | 473-30-3 |
| Analyte or Component Names | Thiazolsulfone |
| Shipping Condition | Room Temperature |
| Molecular Formula | C9 H9 N3 O2 S2 |
| Certifications/Compliance | ISO 17025 |
| Synonym | 2-Thiazolamine, 5-[(4-aminophenyl)sulfonyl]-, 2-Amino-5-sulfanilylthiazole, NSC 1884, Promizol, Promizole, Protozol, Thiazolesulfone, Thiazosulfone |
| InChI Formula | InChI=1S/C9H9N3O2S2/c10-6-1-3-7(4-2-6)16(13,14)8-5-12-9(11)15-8/h1-5H,10H2,(H2,11,12) |
| SMILES | Nc1ccc(cc1)S(=O)(=O)c2cnc(N)s2 |
| IUPAC Name | 5-(4-aminophenyl)sulfonyl-1,3-thiazol-2-amine |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction