Learn More
Valeric Acid Sodium Salt, TRC
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Chemical Name or Material | Valeric acid sodium salt |
| CAS | 6106-41-8 |
| Molecular Formula | C5 H9 O2 . Na |
| Quantity | 100 mg |
| Synonym | Pentanoic acid','sodium salt (1:1), Pentanoic acid','sodium salt (9CI), Valeric acid','sodium salt (8CI), Sodium n-pentanoate, Sodium n-valerate, Sodium pentanoate, Sodium valerate |
| InChI Formula | InChI=1S/C5H10O2.Na/c1-2-3-4-5(6)7;/h2-4H2,1H3,(H,6,7);/q;+1/p-1 |
| SMILES | [Na+].CCCCC(=O)[O-] |
| IUPAC Name | sodium;pentanoate |
| Molecular Weight (g/mol) | 124.11 |
| Formula Weight | 124.05 |