Learn More
Xantphos Pd G3, TRC
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Chemical Name or Material | Xantphos Pd G3 |
| CAS | 1445085-97-1 |
| Molecular Formula | C52H45NO4P2PdS |
| Quantity | 100 mg |
| Synonym | [2'-(Amino-kN)[1,1'-biphenyl]-2-yl-kC][[5-(diphenylphosphino)-9,9-dimethyl-9H-xanthen-4-yl]diphenylphosphine-kP](methanesulfonato-kO)palladium |
| InChI Formula | InChI=1S/C39H32OP2.C12H10N.CH4O3S.Pd/c1-39(2)33-25-15-27-35(41(29-17-7-3-8-18-29)30-19-9-4-10-20-30)37(33)40-38-34(39)26-16-28-36(38)42(31-21-11-5-12-22-31)32-23-13-6-14-24-32;13-12-9-5-4-8-11(12)10-6-2-1-3-7-10;1-5(2,3)4;/h3-28H,1-2H3;1-6,8-9H,13H2;1H3,(H,2,3,4);/q;;;+1/p-1 |
| SMILES | CC1(C)c2cccc(P(c3ccccc3)c4ccccc4)c2Oc5c1cccc5[P](c6ccccc6)(c7ccccc7)[Pd]8(OS(=O)(=O)C)c9ccccc9c%10ccccc%10[NH2]8 |
| Molecular Weight (g/mol) | 948.35 |
| Formula Weight | 947.1579 |