missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(Z)-1-(Diphenylmethyl)-4-(3-phenylprop-2-enyl)piperazine, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
(Z)-1-(Diphenylmethyl)-4-(3-phenylprop-2-enyl)piperazine
Specifications
Specifications
| Chemical Name or Material | (Z)-1-(Diphenylmethyl)-4-(3-phenylprop-2-enyl)piperazine |
| CAS | 750512-44-8 |
| Analyte or Component Names | (Z)-1-(Diphenylmethyl)-4-(3-phenylprop-2-enyl)piperazine |
| Shipping Condition | Room Temperature |
| Molecular Formula | C26 H28 N2 |
| Certifications/Compliance | ISO 17025 |
| Synonym | (Z)-1-(Diphenylmethyl)-4-(3-phenylprop-2-enyl)piperazine, Cinnarizine Imp. B (EP), Cinnarizine Impurity B |
| InChI Formula | InChI=1S/C26H28N2/c1-4-11-23(12-5-1)13-10-18-27-19-21-28(22-20-27)26(24-14-6-2-7-15-24)25-16-8-3-9-17-25/h1-17,26H,18-22H2/b13-10- |
| SMILES | C(\C=C/c1ccccc1)N2CCN(CC2)C(c3ccccc3)c4ccccc4 |
| IUPAC Name | 1-benzhydryl-4-[(Z)-3-phenylprop-2-enyl]piperazine |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction